Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Contour™ Upholstered Lab Chairs

Supplier: VWR International
VWR® Contour™
VSLC-L-1175-R VSLC-M-1175-R VSLC-H-R VSLC-M-133NH-R VSLC-L-R VSLC-M-C133-R VSLC-M-R VSLC-H-1175-R VSLC-L-133NH-R VSLC-H-C133-R VSLC-L-C133-R VSLC-H-133NH-R
Black cloth or vinyl upholstered lab chairs feature a 66 cm (26") diameter composite base with casters, pneumatic seat-height adjustment, backrest tilt, contoured foam cushioned seat, 5.1 cm (2") casters, and a backrest with kidney support.

  • Black cloth or vinyl upholstery
  • Pneumatic seat height adjustment
  • Contoured foam cushioned seat

Medium and high bench height chairs have a 50.8 cm (20") diameter adjustable-height footring.

Ordering information: Contour seating is covered by a five-year warranty against defects in material, workmanship or abnormal wear. Upholstery is covered for one year. Pneumatic cylinders are covered for the life of the product.



 

ORDER

Expand All / Collapse All

Chairs Meet California Technical Bulletin 117 Requirements
Material Seat Height Chair Style Shipping Weight VWR Catalog Number Unit Price Quantity
Cloth 17–22" Desk Height 29 lbs. 97056-539 Each Retrieving
Product available on GSA Advantage®
Cloth 21–28" Medium Bench Height 33 lbs. 97056-541 Each Retrieving
Product available on GSA Advantage®
Cloth 21–31" High Bench Height 34 lbs. 97056-543 Each Retrieving
Product available on GSA Advantage®
Vinyl 17–22" Desk Height 29 lbs. 80086-411 Each Retrieving
Product available on GSA Advantage®
Vinyl 21–28" Medium Bench Height 33 lbs. 80086-413 Each Retrieving
Product available on GSA Advantage®
Vinyl 21–31" High Bench Height 34 lbs. 80086-415 Each Retrieving
Product available on GSA Advantage®

Chairs Meet California Technical Bulletin 117 and 133 Requirements
Material Seat Height Chair Style Shipping Weight VWR Catalog Number Unit Price Quantity
Cloth 17–22" Desk Height 29 lbs. 97056-563 Each Retrieving
Product available on GSA Advantage®
Cloth 21–28" Medium Bench Height 33 lbs. 97056-565 Each Retrieving
Product available on GSA Advantage®
Cloth 21–31" High Bench Height 34 lbs. 97056-567 Each Retrieving
Product available on GSA Advantage®
Vinyl 17–22" Desk Height 29 lbs. 14555-333 Each Retrieving
Product available on GSA Advantage®
Vinyl 21–28" Medium Bench Height 33 lbs. 14555-335 Each Retrieving
Product available on GSA Advantage®
Vinyl 21–31" High Bench Height 34 lbs. 14555-337 Each Retrieving
Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Seat Dimensions 181/2 x 161/2 x 3"
Backrest Dimensions 141/2 x 91/2"

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping