Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ultra PFP (USP L43) 3 µm and 5 µm Columns, Restek

Supplier: RESTEK CORP
9176535 9176513 9176515 9176561 9176563 9176562 9176565 9176521 9176523 9176522 9176525 9176572 9176352 9176351 9176571 9176552 9176332 9176353 9176331 9176573 9176551 9176532 9176355 9176575 9176311 9176553 9176333 9176531 9176512 9176335 9176533 9176555 9176511 9176313
Pentafluorophenyl (PFP) phase columns provide unique selectivity for compounds containing organohalogens or other basic functional groups.

  • Ideal for use with halogenated compounds and amides
  • Fully endcapped

Ultra PFP columns are ideal for taxol and precursor, halogenated compounds, ketones, esters, and amides.




 

ORDER

Expand All / Collapse All
Pore Size Particle Size I.D. x L Phase Column Format Supplier No. VWR Catalog Number Unit Price Quantity
100 Å 5 µm 4.6 x 150 mm Ultra PFP Analytical 9176565 89048-140 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 89048-140
pH Range 2.5 to 7.5
Temperature Limit 80°C (176°F)
Carbon Load 7%

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty