Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

StarRise™ (100Å Pore Size) HPLC Columns, ES Industries

Supplier: ES INDUSTRIES
StarRise™
124221SNRC8 15D22SNRC8 112121SNRC18 124121SNRC8 11D121SNRC8 194121SNRC18 154221SNRC8 13D221SNRC18 122121SNRC8 122221SNRC8 122221SNRC18 115121SNRC18 15222SNRC18 124121SNRC18 19D121SNRC18 194121SNRC8 192121SNRC18 134121SNRC18 192121SNRC8 13D221SNRC8 132121SNRC8 12D121SNRC18 125221SNRC18 114221SNRC8 122121SNRC18 132221SNRC18 115121SNRC8 135221SNRC8 155221SNRC18 134121SNRC8 114221SNRC18 125221SNRC8 124221SNRC18 11D221SNRC18 155221SNRC8 13D121SNRC18 135121SNRC18 112221SNRC18 12D221SNRC8 12D121SNRC8 112221SNRC8 154221SNRC18 115221SNRC18 112121SNRC8 125121SNRC8 132221SNRC8 13D121SNRC8 114121SNRC18 19D121SNRC8 12D221SNRC18 15222SNRC8 134221SNRC18 195121SNRC18 114121SNRC8 132121SNRC18 135221SNRC18 115221SNRC8 125121SNRC18 11D221SNRC8 135121SNRC8 134221SNRC8 195121SNRC8 15D22SNRC18 11D121SNRC18
These columns for reversed phase chromatography are an excellent equivalent to Waters® SunFire™ columns

  • Long lifetime and excellent reproducibility

They provide symmetrical peaks for improved resolution and quantization of acidic, neutral, and basic compounds.




 

ORDER

Expand All / Collapse All
Column Format Phase I.D. x L Packing Material Particle Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical C18 4.0 x 150 mm Silica 3.5 µm 134121SNRC18 101311-606 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 50 mm Silica 3.5 µm 114121SNRC8 101311-678 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 75 mm Silica 3.5 µm 194121SNRC8 101311-686 Each Retrieving
View MSDS SDS
Analytical C8 2.1 x 100 mm Silica 5 µm 122221SNRC8 101311-690 Each Retrieving
View MSDS SDS
Analytical C8 2.1 x 50 mm Silica 3.5 µm 112121SNRC8 101311-674 Each Retrieving
View MSDS SDS
Analytical C18 2.1 x 150 mm Silica 3.5 µm 132121SNRC18 101311-602 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 150 mm Silica 5 µm 134221SNRC8 101311-702 Each Retrieving
View MSDS SDS
Analytical C18 2.1 x 100 mm Silica 5 µm 122221SNRC18 101311-626 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 50 mm Silica 5 µm 114221SNRC8 101311-718 Each Retrieving
View MSDS SDS
Analytical C18 2.1 x 50 mm Silica 3.5 µm 112121SNRC18 101311-610 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 100 mm Silica 5 µm 125221SNRC8 101311-696 Each Retrieving
View MSDS SDS
Analytical C8 2.1 x 150 mm Silica 5 µm 132221SNRC8 101311-698 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 150 mm Silica 5 µm 135221SNRC18 101311-640 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 250 mm Silica 5 µm 154221SNRC18 101311-646 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 150 mm Silica 5 µm 135221SNRC8 101311-704 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 50 mm Silica 5 µm 114221SNRC18 101311-654 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 250 mm Silica 5 µm 155221SNRC8 101311-712 Each Retrieving
View MSDS SDS
Analytical C18 2.1 x 75 mm Silica 3.5 µm 192121SNRC18 101311-618 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 50 mm Silica 5 µm 115221SNRC8 101311-720 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 100 mm Silica 3.5 µm 125121SNRC18 101311-600 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 75 mm Silica 3.5 µm 195121SNRC8 101311-688 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 50 mm Silica 3.5 µm 115121SNRC8 101311-680 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 100 mm Silica 5 µm 124221SNRC8 101311-694 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 100 mm Silica 3.5 µm 125121SNRC8 101311-664 Each Retrieving
View MSDS SDS
Analytical C8 2.1 x 50 mm Silica 5 µm 112221SNRC8 101311-714 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 250 mm Silica 5 µm 154221SNRC8 101311-710 Each Retrieving
View MSDS SDS
Analytical C8 2.1 x 75 mm Silica 3.5 µm 192121SNRC8 101311-682 Each Retrieving
View MSDS SDS
Analytical C18 2.1 x 100 mm Silica 3.5 µm 122121SNRC18 101311-594 Each Retrieving
View MSDS SDS
Analytical C8 2.1 x 150 mm Silica 3.5 µm 132121SNRC8 101311-666 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 100 mm Silica 3.5 µm 124121SNRC18 101311-598 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 150 mm Silica 3.5 µm 135121SNRC18 101311-608 Each Retrieving
View MSDS SDS
Analytical C18 2.1 x 150 mm Silica 5 µm 132221SNRC18 101311-634 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 100 mm Silica 5 µm 125221SNRC18 101311-632 Each Retrieving
View MSDS SDS
Analytical C8 2.1 x 100 mm Silica 3.5 µm 122121SNRC8 101311-658 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 150 mm Silica 3.5 µm 134121SNRC8 101311-670 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 75 mm Silica 3.5 µm 194121SNRC18 101311-622 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 50 mm Silica 3.5 µm 114121SNRC18 101311-614 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 50 mm Silica 5 µm 115221SNRC18 101311-656 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 250 mm Silica 5 µm 155221SNRC18 101311-648 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 100 mm Silica 3.5 µm 124121SNRC8 101311-662 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 150 mm Silica 5 µm 134221SNRC18 101311-638 Each Retrieving
View MSDS SDS
Analytical C18 2.1 x 50 mm Silica 5 µm 112221SNRC18 101311-650 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 100 mm Silica 5 µm 124221SNRC18 101311-630 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 50 mm Silica 3.5 µm 115121SNRC18 101311-616 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 150 mm Silica 3.5 µm 135121SNRC8 101311-672 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 75 mm Silica 3.5 µm 195121SNRC18 101311-624 Each Retrieving
View MSDS SDS
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description C18 C8 C18 C8 C18 C8 C18 C8 C18 C8 C18 C8 C18 C8 C18 C8 C18 C8 C18 C8 C18 C8 C18 C8 C18
Carbon Load 16% 11.5% 16% 11.5% 16% 11.5% 16% 11.5% 16% 11.5% 16% 11.5% 16% 11.5% 16% 11.5% 16% 11.5% 16% 11.5% 16% 11.5% 16% 11.5% 16%
Endcapped Yes

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping