Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sonoma Silica(2) (100Å Pore Size) HPLC Columns, ES Industries

Supplier: ES INDUSTRIES
194221SMASI2 165B21SMASI2 152B2SMASI2 165321SMASI2 11D32SMASI2 124221SMASI2 135B21SMASI2 132B21SMASI2 16232SMASI2 125121SMASI2 11D221SMASI2 134321SMASI2 192121SMASI2 122121SMASI2 13D121SMASI2 122321SMASI2 154221SMASI2 195121SMASI2 125321SMASI2 155221SMASI2 114B21SMASI2 135321SMASI2 132221SMASI2 15222SMASI2 114221SMASI2 125B21SMASI2 134121SMASI2 164321SMASI2 15D32SMASI2 16D32SMASI2 112121SMASI2 115221SMASI2 12D121SMASI2 154B21SMASI2 124321SMASI2 19D121SMASI2 135121SMASI2 12D321SMASI2 112321SMASI2 13DB2SMASI2 194121SMASI2 124B21SMASI2 154321SMASI2 12DB2SMASI2 112B21SMASI2 124121SMASI2 115B21SMASI2 11D121SMASI2 115121SMASI2 15DB2SMASI2 19D22SMASI2 192221SMASI2 195221SMASI2 125221SMASI2 162B2SMASI2 134221SMASI2 155B21SMASI2 122221SMASI2 13D221SMASI2 164B21SMASI2 132321SMASI2 114121SMASI2 15232SMASI2 11DB2SMASI2 155321SMASI2 15D22SMASI2 114321SMASI2 134B21SMASI2 16DB2SMASI2 13D32SMASI2 112221SMASI2 115321SMASI2 12D221SMASI2 122B21SMASI2 132121SMASI2 135221SMASI2
These columns are an excellent equivalent to Phenomenex Luna® Silica(2) columns

Columns are 99.999% pure and have enhanced surface smoothness and sphericity.




 

ORDER

Expand All / Collapse All
Column Format Phase I.D. x L Packing Material Particle Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical Silica 4.6 x 150 mm Silica 5 µm 135221SMASI2 101310-112 Each Retrieving
View MSDS SDS
Analytical Silica 2.1 x 150 mm Silica 3 µm 132121SMASI2 101310-074 Each Retrieving
View MSDS SDS
Analytical Silica 4.6 x 250 mm Silica 15 µm 155B21SMASI2 101310-048 Each Retrieving
View MSDS SDS
Analytical Silica 3.2 x 150 mm Silica 3 µm 13D121SMASI2 101310-076 Each Retrieving
View MSDS SDS
Analytical Silica 2.1 x 50 mm Silica 3 µm 112121SMASI2 101310-082 Each Retrieving
View MSDS SDS
Analytical Silica 4.0 x 150 mm Silica 5 µm 134221SMASI2 101310-110 Each Retrieving
View MSDS SDS
Analytical Silica 4.6 x 150 mm Silica 15 µm 135B21SMASI2 101310-040 Each Retrieving
View MSDS SDS
Analytical Silica 2.1 x 100 mm Silica 5 µm 122221SMASI2 101310-098 Each Retrieving
View MSDS SDS
Analytical Silica 2.1 x 150 mm Silica 5 µm 132221SMASI2 101310-106 Each Retrieving
View MSDS SDS
Analytical Silica 4.6 x 300 mm Silica 15 µm 165B21SMASI2 101310-056 Each Retrieving
View MSDS SDS
Analytical Silica 3.2 x 50 mm Silica 3 µm 11D121SMASI2 101310-084 Each Retrieving
View MSDS SDS
Analytical Silica 2.1 x 75 mm Silica 3 µm 192121SMASI2 101310-090 Each Retrieving
View MSDS SDS
Analytical Silica 4.6 x 50 mm Silica 3 µm 115121SMASI2 101310-088 Each Retrieving
View MSDS SDS
Analytical Silica 2.1 x 100 mm Silica 3 µm 122121SMASI2 101310-066 Each Retrieving
View MSDS SDS
Analytical Silica 4.0 x 50 mm Silica 3 µm 114121SMASI2 101310-086 Each Retrieving
View MSDS SDS
Analytical Silica 4.6 x 100 mm Silica 3 µm 125121SMASI2 101310-072 Each Retrieving
View MSDS SDS
Analytical Silica 3.2 x 75 mm Silica 3 µm 19D121SMASI2 101310-092 Each Retrieving
View MSDS SDS
Analytical Silica 4.6 x 250 mm Silica 5 µm 155221SMASI2 101310-120 Each Retrieving
View MSDS SDS
Analytical Silica 4.0 x 100 mm Silica 3 µm 124121SMASI2 101310-070 Each Retrieving
View MSDS SDS
Analytical Silica 4.6 x 50 mm Silica 5 µm 115221SMASI2 101310-128 Each Retrieving
View MSDS SDS
Analytical Silica 4.0 x 75 mm Silica 3 µm 194121SMASI2 101310-094 Each Retrieving
View MSDS SDS
Analytical Silica 4.6 x 100 mm Silica 5 µm 125221SMASI2 101310-104 Each Retrieving
View MSDS SDS
Analytical Silica 4.0 x 75 mm Silica 5 µm 194221SMASI2 101310-134 Each Retrieving
View MSDS SDS
Analytical Silica 3.2 x 50 mm Silica 5 µm 11D221SMASI2 101310-124 Each Retrieving
View MSDS SDS
Analytical Silica 4.6 x 300 mm Silica 10 µm 165321SMASI2 101310-016 Each Retrieving
View MSDS SDS
Analytical Silica 2.1 x 75 mm Silica 5 µm 192221SMASI2 101310-130 Each Retrieving
View MSDS SDS
Analytical Silica 4.6 x 75 mm Silica 5 µm 195221SMASI2 101310-136 Each Retrieving
View MSDS SDS
Analytical Silica 4.0 x 100 mm Silica 5 µm 124221SMASI2 101310-102 Each Retrieving
View MSDS SDS
Analytical Silica 4.6 x 75 mm Silica 3 µm 195121SMASI2 101310-096 Each Retrieving
View MSDS SDS
Analytical Silica 4.6 x 150 mm Silica 3 µm 135121SMASI2 101310-080 Each Retrieving
View MSDS SDS
Analytical Silica 3.2 x 100 mm Silica 5 µm 12D221SMASI2 101310-100 Each Retrieving
View MSDS SDS
Analytical Silica 3.2 x 150 mm Silica 5 µm 13D221SMASI2 101310-108 Each Retrieving
View MSDS SDS
Analytical Silica 3.2 x 100 mm Silica 3 µm 12D121SMASI2 101310-068 Each Retrieving
View MSDS SDS
Analytical Silica 4.0 x 150 mm Silica 3 µm 134121SMASI2 101310-078 Each Retrieving
View MSDS SDS
Analytical Silica 4.6 x 250 mm Silica 10 µm 155321SMASI2 101310-008 Each Retrieving
View MSDS SDS
Analytical Silica 4.0 x 250 mm Silica 5 µm 154221SMASI2 101310-118 Each Retrieving
View MSDS SDS
Analytical Silica 4.0 x 50 mm Silica 5 µm 114221SMASI2 101310-126 Each Retrieving
View MSDS SDS
Analytical Silica 2.1 x 50 mm Silica 5 µm 112221SMASI2 101310-122 Each Retrieving
View MSDS SDS
Analytical Silica 4.6 x 150 mm Silica 10 µm 135321SMASI2 101310-000 Each Retrieving
View MSDS SDS
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

pH Range 1. 5 - 10
Surface Area 400 m²/g
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping