Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Neptune dC18 (100Å Pore Size) HPLC Columns, ES Industries

Supplier: ES INDUSTRIES
135221NPNDC18 154221NPNDC18 122221NPNDC18 124221NPNDC18 125121NPNDC18 112121NPNDC18 134121NPNDC18 125221NPNDC18 194221NPNDC18 112221NPNDC18 114221NPNDC18 195121NPNDC18 192221NPNDC18 115121NPNDC18 124121NPNDC18 134221NPNDC18 115221NPNDC18 155221NPNDC18 192121NPNDC18 132121NPNDC18 195221NPNDC18 114121NPNDC18 132221NPNDC18 135121NPNDC18 194121NPNDC18 122121NPNDC18
These columns for reversed phase chromatography are an excellent equivalent to Waters® Atlantis® dC18 columns

  • Long lifetime and excellent reproducibility

They feature enhanced polar compound retention with complete compatibilty with aqueous mobile phases.




 

ORDER

Expand All / Collapse All
Column Format I.D. x L Particle Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical 4.0 x 75 mm 3 µm 194121NPNDC18 101308-928 Each Retrieving
View MSDS SDS
Analytical 4.0 x 50 mm 3 µm 114121NPNDC18 101308-920 Each Retrieving
View MSDS SDS
Analytical 4.6 x 100 mm 5 µm 125221NPNDC18 101308-938 Each Retrieving
View MSDS SDS
Analytical 4.6 x 150 mm 5 µm 135221NPNDC18 101308-946 Each Retrieving
View MSDS SDS
Analytical 4.6 x 100 mm 3 µm 125121NPNDC18 101308-906 Each Retrieving
View MSDS SDS
Analytical 4.0 x 250 mm 5 µm 154221NPNDC18 101308-952 Each Retrieving
View MSDS SDS
Analytical 4.6 x 250 mm 5 µm 155221NPNDC18 101308-954 Each Retrieving
View MSDS SDS
Analytical 2.1 x 50 mm 3 µm 112121NPNDC18 101308-916 Each Retrieving
View MSDS SDS
Analytical 2.1 x 150 mm 5 µm 132221NPNDC18 101308-940 Each Retrieving
View MSDS SDS
Analytical 4.6 x 75 mm 3 µm 195121NPNDC18 101308-930 Each Retrieving
View MSDS SDS
Analytical 2.1 x 100 mm 5 µm 122221NPNDC18 101308-932 Each Retrieving
View MSDS SDS
Analytical 4.0 x 150 mm 5 µm 134221NPNDC18 101308-944 Each Retrieving
View MSDS SDS
Analytical 4.0 x 150 mm 3 µm 134121NPNDC18 101308-912 Each Retrieving
View MSDS SDS
Analytical 4.0 x 100 mm 5 µm 124221NPNDC18 101308-936 Each Retrieving
View MSDS SDS
Analytical 4.6 x 50 mm 3 µm 115121NPNDC18 101308-922 Each Retrieving
View MSDS SDS
Analytical 4.6 x 150 mm 3 µm 135121NPNDC18 101308-914 Each Retrieving
View MSDS SDS
Analytical 2.1 x 150 mm 3 µm 132121NPNDC18 101308-908 Each Retrieving
View MSDS SDS
Analytical 4.0 x 75 mm 5 µm 194221NPNDC18 101308-968 Each Retrieving
View MSDS SDS
Analytical 4.0 x 50 mm 5 µm 114221NPNDC18 101308-960 Each Retrieving
View MSDS SDS
Analytical 4.6 x 75 mm 5 µm 195221NPNDC18 101308-970 Each Retrieving
View MSDS SDS
Analytical 2.1 x 75 mm 3 µm 192121NPNDC18 101308-924 Each Retrieving
View MSDS SDS
Analytical 2.1 x 50 mm 5 µm 112221NPNDC18 101308-956 Each Retrieving
View MSDS SDS
Analytical 2.1 x 75 mm 5 µm 192221NPNDC18 101308-964 Each Retrieving
View MSDS SDS
Analytical 4.6 x 50 mm 5 µm 115221NPNDC18 101308-962 Each Retrieving
View MSDS SDS
Analytical 2.1 x 100 mm 3 µm 122121NPNDC18 101308-900 Each Retrieving
View MSDS SDS
Analytical 4.0 x 100 mm 3 µm 124121NPNDC18 101308-904 Each Retrieving
View MSDS SDS
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping