Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Epic Silica HILIC HPLC Columns, 120 Å, ES Industries

Supplier: ES INDUSTRIES
Epic®
122191ESI 132191ESI 112191ESI 135291ESI 114191ESI 155291ESI 125291ESI 115291ESI 11D291ESI 112291ESI 124191ESI 134191ESI 132291ESI 122291ESI 13D291ESI 12D291ESI 15D291ESI 124291ESI 13D191ESI 152291ESI 11D191ESI 134291ESI 114291ESI 12D191ESI 154291ESI 125191ESI 115191ESI 135191ESI
Reversed-phase HPLC is widely used for separation of many pharmaceutical compounds

However, retention and separation of many polar analytes has proven to be a challenge. Many of these types of compounds are unretained or poorly retained on most conventional reversed-phase stationary phases, such as ODS. Fortunately, to deal with these types of analytes an alternative mode of chromatography can be utilized using polar stationary phases and highly organic mobile phases. This mode of chromatography has been utilized for many years and is referred to as “hydrophilic interaction chromatography” or HILIC. HILIC chromatography uses mobile phases containing between 5 - 20 % water for the retention of polar compounds. ES Industries is pleased to offer a number of stationary phases based traditional bonded phases such as amine and diol. The columns are produced from ultra high purity silica using the most advanced bonding technology available. In addition, the columns are packed to the highest possible standards yielding superior performance and durability.




 

ORDER

Expand All / Collapse All
Column Format Phase I.D. x L Packing Material Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical Silica HILIC 4.6 x 250 mm Silica 5 µm 120 Å 155291ESI 95007-464 Each Retrieving
View MSDS SDS
Analytical Silica HILIC 4.6 x 50 mm Silica 3 µm 120 Å 115191ESI 95007-416 Each Retrieving
View MSDS SDS
Analytical Silica HILIC 4.6 x 100 mm Silica 3 µm 120 Å 125191ESI 95007-424 Each Retrieving
View MSDS SDS
Analytical Silica HILIC 4.6 x 150 mm Silica 3 µm 120 Å 135191ESI 95007-432 Each Retrieving
View MSDS SDS
Analytical Silica HILIC 4.6 x 100 mm Silica 5 µm 120 Å 125291ESI 95007-448 Each Retrieving
View MSDS SDS
Analytical Silica HILIC 4.6 x 150 mm Silica 5 µm 120 Å 135291ESI 95007-456 Each Retrieving
View MSDS SDS
Analytical Silica HILIC 4.6 x 50 mm Silica 5 µm 120 Å 115291ESI 95007-440 Each Retrieving
View MSDS SDS
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping