Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Epic Phenyl Hexyl HPLC Columns, 120 Å, ES Industries

Supplier: ES INDUSTRIES
Epic®
11D191EPHX 122291EPHX 114291EPHX 112291EPHX 12D191EPHX 115291EPHX 125291EPHX 124291EPHX 13D191EPHX 134291EPHX 132291EPHX 135291EPHX 132191EPHX 115191EPHX 125191EPHX 124191EPHX 134191EPHX 135191EPHX 11D291EPHX 12D291EPHX 13D291EPHX 154291EPHX 155291EPHX 152291EPHX 112191EPHX 122191EPHX 15D291EPHX 114191EPHX
Epic Phenyl-Hexyl uses a hexyl-linked phenyl phase

The hexyl alkyl chain provides for unique selectivity and increased hydrolytic stability when compared to propyl-linked chemistry. The Epic high-density bonding technology delivers to the chromatographer superior performance, durability, and enhanced lot-to-lot reproducibility. Epic Phenyl-Hexyl offers the chromatographer a truly superior phenyl based interaction for enhanced chromatographic performance.




 

ORDER

Expand All / Collapse All
Column Format Phase I.D. x L Packing Material Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical Phenyl-Hexyl 4.6 x 100 mm Silica 3 µm 120 Å 125191EPHX 95007-368 Each Retrieving
View MSDS SDS
Analytical Phenyl-Hexyl 4.6 x 150 mm Silica 3 µm 120 Å 135191EPHX 95007-376 Each Retrieving
View MSDS SDS
Analytical Phenyl-Hexyl 4.6 x 50 mm Silica 3 µm 120 Å 115191EPHX 95007-360 Each Retrieving
View MSDS SDS
Analytical Phenyl-Hexyl 4.6 x 50 mm Silica 5 µm 120 Å 115291EPHX 95007-384 Each Retrieving
View MSDS SDS
Analytical Phenyl-Hexyl 4.6 x 100 mm Silica 5 µm 120 Å 125291EPHX 95007-392 Each Retrieving
View MSDS SDS
Analytical Phenyl-Hexyl 4.6 x 150 mm Silica 5 µm 120 Å 135291EPHX 95007-400 Each Retrieving
View MSDS SDS
Analytical Phenyl-Hexyl 4.6 x 250 mm Silica 5 µm 120 Å 155291EPHX 95007-408 Each Retrieving
View MSDS SDS
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty