Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sonoma CN (100Å Pore Size) HPLC Columns, ES Industries

Supplier: ES INDUSTRIES
195121SMACN 135121SMACN 165321SMACN 16D32SMACN 135321SMACN 19D121SMACN 135221SMACN 19D221SMACN 114221SMACN 114321SMACN 195221SMACN 15232SMACN 11D121SMACN 112121SMACN 114121SMACN 112321SMACN 11D321SMACN 122321SMACN 122121SMACN 154221SMACN 124321SMACN 13D221SMACN 125221SMACN 124121SMACN 155321SMACN 134221SMACN 194221SMACN 194121SMACN 134121SMACN 134321SMACN 192221SMACN 192121SMACN 12D321SMACN 115221SMACN 115321SMACN 115121SMACN 12D221SMACN 12D121SMACN 16232SMACN 132221SMACN 132121SMACN 15D32SMACN 132321SMACN 164321SMACN 11D221SMACN 112221SMACN 122221SMACN 15222SMACN 154321SMACN 13D321SMACN 155221SMACN 15D22SMACN 125121SMACN 125321SMACN 13D121SMACN 124221SMACN
These columns are an excellent equivalent to Phenomenex Luna® CN columns

They deliver excellent efficiency, peak shape, and resolution for virtually all HPLC applications. The range of particle sizes offers versatility for capillary and LC/MS to process applications.




 

ORDER

Expand All / Collapse All
Column Format Phase I.D. x L Packing Material Particle Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical Cyano 2.1 x 75 mm Silica 3 µm 192121SMACN 101309-786 Each Retrieving
View MSDS SDS
Analytical Cyano 4.0 x 100 mm Silica 5 µm 124221SMACN 101309-798 Each Retrieving
View MSDS SDS
Analytical Cyano 4.0 x 150 mm Silica 5 µm 134221SMACN 101309-806 Each Retrieving
View MSDS SDS
Analytical Cyano 4.0 x 250 mm Silica 5 µm 154221SMACN 101309-814 Each Retrieving
View MSDS SDS
Analytical Cyano 2.1 x 50 mm Silica 5 µm 112221SMACN 101309-818 Each Retrieving
View MSDS SDS
Analytical Cyano 3.2 x 50 mm Silica 5 µm 11D221SMACN 101309-820 Each Retrieving
View MSDS SDS
Analytical Cyano 4.0 x 50 mm Silica 5 µm 114221SMACN 101309-822 Each Retrieving
View MSDS SDS
Analytical Cyano 3.2 x 75 mm Silica 5 µm 19D221SMACN 101309-828 Each Retrieving
View MSDS SDS
Analytical Cyano 4.0 x 75 mm Silica 5 µm 194221SMACN 101309-830 Each Retrieving
View MSDS SDS
Analytical Cyano 4.6 x 75 mm Silica 5 µm 195221SMACN 101309-832 Each Retrieving
View MSDS SDS
Analytical Cyano 4.0 x 50 mm Silica 3 µm 114121SMACN 101309-782 Each Retrieving
View MSDS SDS
Analytical Cyano 4.6 x 75 mm Silica 3 µm 195121SMACN 101309-792 Each Retrieving
View MSDS SDS
Analytical Cyano 4.6 x 50 mm Silica 5 µm 115221SMACN 101309-824 Each Retrieving
View MSDS SDS
Analytical Cyano 4.0 x 300 mm Silica 10 µm 164321SMACN 101309-750 Each Retrieving
View MSDS SDS
Analytical Cyano 4.0 x 150 mm Silica 3 µm 134121SMACN 101309-774 Each Retrieving
View MSDS SDS
Analytical Cyano 3.2 x 75 mm Silica 3 µm 19D121SMACN 101309-788 Each Retrieving
View MSDS SDS
Analytical Cyano 4.0 x 75 mm Silica 3 µm 194121SMACN 101309-790 Each Retrieving
View MSDS SDS
Analytical Cyano 2.1 x 100 mm Silica 5 µm 122221SMACN 101309-794 Each Retrieving
View MSDS SDS
Analytical Cyano 2.1 x 150 mm Silica 5 µm 132221SMACN 101309-802 Each Retrieving
View MSDS SDS
Analytical Cyano 3.2 x 150 mm Silica 5 µm 13D221SMACN 101309-804 Each Retrieving
View MSDS SDS
Analytical Cyano 2.1 x 75 mm Silica 5 µm 192221SMACN 101309-826 Each Retrieving
View MSDS SDS
Analytical Cyano 3.2 x 100 mm Silica 5 µm 12D221SMACN 101309-796 Each Retrieving
View MSDS SDS
Analytical Cyano 4.6 x 250 mm Silica 10 µm 155321SMACN 101309-744 Each Retrieving
View MSDS SDS
Analytical Cyano 4.6 x 150 mm Silica 3 µm 135121SMACN 101309-776 Each Retrieving
View MSDS SDS
Analytical Cyano 4.6 x 100 mm Silica 3 µm 125121SMACN 101309-768 Each Retrieving
View MSDS SDS
Analytical Cyano 4.6 x 150 mm Silica 5 µm 135221SMACN 101309-808 Each Retrieving
View MSDS SDS
Analytical Cyano 4.6 x 250 mm Silica 5 µm 155221SMACN 101309-816 Each Retrieving
View MSDS SDS
Analytical Cyano 4.0 x 100 mm Silica 3 µm 124121SMACN 101309-766 Each Retrieving
View MSDS SDS
Analytical Cyano 4.6 x 150 mm Silica 10 µm 135321SMACN 101309-736 Each Retrieving
View MSDS SDS
Analytical Cyano 4.6 x 300 mm Silica 10 µm 165321SMACN 101309-752 Each Retrieving
View MSDS SDS
Analytical Cyano 3.2 x 100 mm Silica 3 µm 12D121SMACN 101309-764 Each Retrieving
View MSDS SDS
Analytical Cyano 3.2 x 150 mm Silica 3 µm 13D121SMACN 101309-772 Each Retrieving
View MSDS SDS
Analytical Cyano 3.2 x 50 mm Silica 3 µm 11D121SMACN 101309-780 Each Retrieving
View MSDS SDS
Analytical Cyano 4.6 x 100 mm Silica 5 µm 125221SMACN 101309-800 Each Retrieving
View MSDS SDS
Analytical Cyano 4.0 x 150 mm Silica 10 µm 134321SMACN 101309-734 Each Retrieving
View MSDS SDS
Analytical Cyano 4.0 x 250 mm Silica 10 µm 154321SMACN 101309-742 Each Retrieving
View MSDS SDS
Analytical Cyano 2.1 x 100 mm Silica 3 µm 122121SMACN 101309-762 Each Retrieving
View MSDS SDS
Analytical Cyano 2.1 x 150 mm Silica 3 µm 132121SMACN 101309-770 Each Retrieving
View MSDS SDS
Analytical Cyano 2.1 x 50 mm Silica 3 µm 112121SMACN 101309-778 Each Retrieving
View MSDS SDS
Analytical Cyano 4.6 x 50 mm Silica 3 µm 115121SMACN 101309-784 Each Retrieving
View MSDS SDS
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Carbon Load 17.5%
pH Range 1.5 - 10
Surface Area 400 m²/g

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty