Sonoma C5 HPLC Columns, 100 Å, ES Industries
Supplier: ES Industries
16532SMAC5 13522SMAC5 15422SMAC5 16432SMAC5 11422SMAC5 15522SMAC5 13422SMAC5 12422SMAC5 12522SMAC5 11522SMAC5 15532SMAC5 13532SMAC5 19422SMAC5 11222SMAC5 13222SMAC5 13D22SMAC5 15432SMAC5 13432SMAC5 12222SMAC5 11D22SMAC5 12D22SMAC5 19522SMAC5
101309-518EA 1191.54 USD
101309-518 101309-534 101309-512 101309-556 101309-544 101309-566 101309-554 101309-510 101309-532 101309-520 101309-542 101309-538 101309-504 101309-558 101309-536 101309-502 101309-568 101309-552 101309-530 101309-540 101309-550 101309-560
Sonoma C5 HPLC Columns, 100 Å, ES Industries
Chromatography Columns
These columns are an excellent equivalent to Phenomenex Luna® C5 columns.
They deliver excellent efficiency, peak shape, and resolution for virtually all HPLC applications. The range of particle sizes offers versatility for capillary and LC/MS to process applications.