Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sonoma C8 (100Å Pore Size) HPLC Columns, ES Industries

Supplier: ES INDUSTRIES
132221SMAC82 114221SMAC82 15232SMAC82 134121SMAC82 134B21SMAC82 15222SMAC82 164321SMAC82 125B21SMAC82 155321SMAC82 13DB2SMAC82 16DB2SMAC82 12D121SMAC82 115321SMAC82 122B21SMAC82 112121SMAC82 16D32SMAC82 124321SMAC82 135221SMAC82 19D121SMAC82 12D321SMAC82 112321SMAC82 124B21SMAC82 194221SMAC82 165B21SMAC82 12DB2SMAC82 112B21SMAC82 124221SMAC82 115121SMAC82 16232SMAC82 13D121SMAC82 134321SMAC82 195221SMAC82 122121SMAC82 11D221SMAC82 125221SMAC82 135B21SMAC82 13D321SMAC82 122321SMAC82 192121SMAC82 154221SMAC82 164B21SMAC82 155221SMAC82 114B21SMAC82 135321SMAC82 114121SMAC82 114321SMAC82 15D32SMAC82 132321SMAC82 15D22SMAC82 11DB2SMAC82 115221SMAC82 12D221SMAC82 112221SMAC82 135121SMAC82 154B21SMAC82 132121SMAC82 19D221SMAC82 194121SMAC82 165321SMAC82 154321SMAC82 11D32SMAC82 11D12SMAC82 124121SMAC82 115B21SMAC82 15DB2SMAC82 152B2SMAC82 132B21SMAC82 125121SMAC82 192221SMAC82 162B2SMAC82 134221SMAC82 125321SMAC82 155B21SMAC82 13D221SMAC82 195121SMAC82 122221SMAC82
These columns are an excellent equivalent to Phenomenex Luna® C8 columns

They deliver excellent efficiency, peak shape, and resolution for virtually all HPLC applications. The range of particle sizes offers versatility for capillary and LC/MS to process applications.




 

ORDER

Expand All / Collapse All
Column Format Phase I.D. x L Packing Material Particle Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical C8 4.6 x 100 mm Silica 5 µm 125221SMAC82 101309-688 Each Retrieving
View MSDS SDS
Analytical C8 2.1 x 150 mm Silica 5 µm 132221SMAC82 101309-690 Each Retrieving
View MSDS SDS
Analytical C8 3.2 x 150 mm Silica 5 µm 13D221SMAC82 101309-692 Each Retrieving
View MSDS SDS
Analytical C8 3.2 x 100 mm Silica 5 µm 12D221SMAC82 101309-684 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 150 mm Silica 3 µm 135121SMAC82 101309-664 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 300 mm Silica 15 µm 164B21SMAC82 101309-638 Each Retrieving
View MSDS SDS
Analytical C8 2.1 x 50 mm Silica 3 µm 112121SMAC82 101309-666 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 150 mm Silica 15 µm 134B21SMAC82 101309-622 Each Retrieving
View MSDS SDS
Analytical C8 2.1 x 100 mm Silica 3 µm 122121SMAC82 101309-650 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 75 mm Silica 5 µm 195221SMAC82 101309-720 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 250 mm Silica 15 µm 155B21SMAC82 101309-632 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 50 mm Silica 5 µm 114221SMAC82 101309-710 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 250 mm Silica 15 µm 154B21SMAC82 101309-630 Each Retrieving
View MSDS SDS
Analytical C8 2.1 x 150 mm Silica 3 µm 132121SMAC82 101309-658 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 150 mm Silica 15 µm 135B21SMAC82 101309-624 Each Retrieving
View MSDS SDS
Analytical C8 3.2 x 100 mm Silica 3 µm 12D121SMAC82 101309-652 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 75 mm Silica 3 µm 195121SMAC82 101309-680 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 50 mm Silica 5 µm 115221SMAC82 101309-712 Each Retrieving
View MSDS SDS
Analytical C8 2.1 x 75 mm Silica 3 µm 192121SMAC82 101309-674 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 250 mm Silica 5 µm 154221SMAC82 101309-702 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 100 mm Silica 3 µm 125121SMAC82 101309-656 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 100 mm Silica 5 µm 124221SMAC82 101309-686 Each Retrieving
View MSDS SDS
Analytical C8 2.1 x 75 mm Silica 5 µm 192221SMAC82 101309-714 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 150 mm Silica 5 µm 134221SMAC82 101309-694 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 300 mm Silica 15 µm 165B21SMAC82 101309-640 Each Retrieving
View MSDS SDS
Analytical C8 3.2 x 75 mm Silica 3 µm 19D121SMAC82 101309-676 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 250 mm Silica 5 µm 155221SMAC82 101309-704 Each Retrieving
View MSDS SDS
Analytical C8 3.2 x 150 mm Silica 3 µm 13D121SMAC82 101309-660 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 150 mm Silica 5 µm 135221SMAC82 101309-696 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 150 mm Silica 10 µm 134321SMAC82 101309-582 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 150 mm Silica 10 µm 135321SMAC82 101309-584 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 50 mm Silica 3 µm 114121SMAC82 101309-670 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 100 mm Silica 3 µm 124121SMAC82 101309-654 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 150 mm Silica 3 µm 134121SMAC82 101309-662 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 50 mm Silica 3 µm 115121SMAC82 101309-672 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 75 mm Silica 3 µm 194121SMAC82 101309-678 Each Retrieving
View MSDS SDS
Analytical C8 3.2 x 50 mm Silica 5 µm 11D221SMAC82 101309-708 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 250 mm Silica 10 µm 154321SMAC82 101309-590 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 300 mm Silica 10 µm 164321SMAC82 101309-598 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 250 mm Silica 10 µm 155321SMAC82 101309-592 Each Retrieving
View MSDS SDS
Analytical C8 2.1 x 100 mm Silica 5 µm 122221SMAC82 101309-682 Each Retrieving
View MSDS SDS
Analytical C8 2.1 x 50 mm Silica 5 µm 112221SMAC82 101309-706 Each Retrieving
View MSDS SDS
Analytical C8 3.2 x 75 mm Silica 5 µm 19D221SMAC82 101309-716 Each Retrieving
View MSDS SDS
Analytical C8 4.6 x 300 mm Silica 10 µm 165321SMAC82 101309-600 Each Retrieving
View MSDS SDS
Analytical C8 4.0 x 75 mm Silica 5 µm 194221SMAC82 101309-718 Each Retrieving
View MSDS SDS
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Carbon Load 17.5%
pH Range 1.5 - 10
Surface Area 400 m²/g

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping