Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sonoma C18 (100Å Pore Size) HPLC Columns, ES Industries

Supplier: ES INDUSTRIES
112B21SMAC182 12222SMAC182 19412SMAC182 13422SMAC182 12522SMAC182 12D22SMAC182 11D12SMAC182 11432SMAC182 135B21SMAC182 19D22SMAC182 164B21SMAC182 11412SMAC182 15522SMAC182 124B21SMAC182 13222SMAC182 13532SMAC182 13D22SMAC182 15D32SMAC182 15222SMAC182 11232SMAC182 15DB2SMAC182 122B21SMAC182 11522SMAC182 11212SMAC182 154B21SMAC182 13512SMAC182 12432SMAC182 11D32SMAC182 134B21SMAC182 115B21SMAC182 19512SMAC182 12412SMAC182 12D12SMAC182 132321SMAC182 11DB2SMAC182 19222SMAC182 15432SMAC182 12232SMAC182 125B21SMAC182 162B2SMAC182 15422SMAC182 12D32SMAC182 12532SMAC182 155B21SMAC182 12212SMAC182 16D32SMAC182 12512SMAC182 13432SMAC182 16532SMAC182 19422SMAC182 114B21SMAC182 13412SMAC182 19D12SMAC182 16232SMAC182 11D22SMAC182 12DB2SMAC182 15532SMAC182 132B21SMAC182 11422SMAC182 13D32SMAC182 16432SMAC182 13522SMAC182 13212SMAC182 11222SMAC182 11532SMAC182 15232SMAC182 13D12SMAC182 12422SMAC182 19212SMAC182 13DB2SMAC182 19522SMAC182 165B21SMAC182 11512SMAC182 16DB2SMAC182 15D22SMAC182 152B2SMAC182
These columns are an excellent equivalent to Phenomenex Luna® C18 columns

They deliver excellent efficiency, peak shape, and resolution for virtually all HPLC applications. The range of particle sizes offers versatility for capillary and LC/MS to process applications.




 

ORDER

Expand All / Collapse All
Column Format Phase I.D. x L Packing Material Particle Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical C18 4.6 x 250 mm Silica 15 µm 155B21SMAC182 101309-400 Each Retrieving
View MSDS SDS
Analytical C18 3.2 x 150 mm Silica 3 µm 13D12SMAC182 101309-428 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 100 mm Silica 3 µm 12512SMAC182 101309-424 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 150 mm Silica 3 µm 13512SMAC182 101309-432 Each Retrieving
View MSDS SDS
Analytical C18 3.2 x 75 mm Silica 3 µm 19D12SMAC182 101309-444 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 75 mm Silica 5 µm 19522SMAC182 101309-488 Each Retrieving
View MSDS SDS
Analytical C18 2.1 x 50 mm Silica 5 µm 11222SMAC182 101309-474 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 75 mm Silica 5 µm 19422SMAC182 101309-486 Each Retrieving
View MSDS SDS
Analytical C18 3.2 x 50 mm Silica 5 µm 11D22SMAC182 101309-476 Each Retrieving
View MSDS SDS
Analytical C18 2.1 x 150 mm Silica 3 µm 13212SMAC182 101309-426 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 100 mm Silica 5 µm 12422SMAC182 101309-454 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 75 mm Silica 3 µm 19512SMAC182 101309-448 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 75 mm Silica 3 µm 19412SMAC182 101309-446 Each Retrieving
View MSDS SDS
Analytical C18 2.1 x 50 mm Silica 3 µm 11212SMAC182 101309-434 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 150 mm Silica 10 µm 13532SMAC182 101309-352 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 250 mm Silica 5 µm 15522SMAC182 101309-472 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 50 mm Silica 3 µm 11412SMAC182 101309-438 Each Retrieving
View MSDS SDS
Analytical C18 2.1 x 75 mm Silica 3 µm 19212SMAC182 101309-442 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 150 mm Silica 5 µm 13522SMAC182 101309-464 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 150 mm Silica 5 µm 13422SMAC182 101309-462 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 150 mm Silica 3 µm 13412SMAC182 101309-430 Each Retrieving
View MSDS SDS
Analytical C18 2.1 x 150 mm Silica 5 µm 13222SMAC182 101309-458 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 50 mm Silica 5 µm 11522SMAC182 101309-480 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 100 mm Silica 5 µm 12522SMAC182 101309-456 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 50 mm Silica 5 µm 11422SMAC182 101309-478 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 300 mm Silica 15 µm 165B21SMAC182 101309-408 Each Retrieving
View MSDS SDS
Analytical C18 2.1 x 100 mm Silica 3 µm 12212SMAC182 101309-418 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 150 mm Silica 10 µm 13432SMAC182 101309-350 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 250 mm Silica 10 µm 15532SMAC182 101309-360 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 150 mm Silica 15 µm 135B21SMAC182 101309-392 Each Retrieving
View MSDS SDS
Analytical C18 2.1 x 50 mm Silica 15 µm 112B21SMAC182 101309-410 Each Retrieving
View MSDS SDS
Analytical C18 3.2 x 50 mm Silica 3 µm 11D12SMAC182 101309-436 Each Retrieving
View MSDS SDS
Analytical C18 3.2 x 100 mm Silica 5 µm 12D22SMAC182 101309-452 Each Retrieving
View MSDS SDS
Analytical C18 3.2 x 150 mm Silica 5 µm 13D22SMAC182 101309-460 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 250 mm Silica 5 µm 15422SMAC182 101309-470 Each Retrieving
View MSDS SDS
Analytical C18 3.2 x 100 mm Silica 3 µm 12D12SMAC182 101309-420 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 100 mm Silica 3 µm 12412SMAC182 101309-422 Each Retrieving
View MSDS SDS
Analytical C18 2.1 x 100 mm Silica 5 µm 12222SMAC182 101309-450 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 50 mm Silica 3 µm 11512SMAC182 101309-440 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 250 mm Silica 10 µm 15432SMAC182 101309-358 Each Retrieving
View MSDS SDS
Analytical C18 4.0 x 300 mm Silica 10 µm 16432SMAC182 101309-366 Each Retrieving
View MSDS SDS
Analytical C18 4.6 x 300 mm Silica 10 µm 16532SMAC182 101309-368 Each Retrieving
View MSDS SDS
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Carbon Load 17.5%
pH Range 1.5 - 10
Surface Area 400 m²/g

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping