Would you like to log in again?
Uvex® Eyewear Cases, Honeywell Safety
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
Uvex®
S487 S490
Cases protect lenses against scratches and abrasions
The economy vinyl case has a top closing flap and strap attachment, and holds all flat-fold spectacles. The nylon bag case features a rip-stop drawstring, and will carry most Uvex® eyewear products. The hard case is equipped with a secure snap closure.
Ordering information: The economy vinyl case is not suitable for Uvex Astro OTG™ 3001 eyewear (see 33000-506 series). The hard case is not suitable for Uvex Astrospec 3000® eyewear (see 33002-036 series).
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Nylon Bag with Rip-Stop Closure S487 14233-345 Each Retrieving
Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty