Would you like to log in again?
Reversed Phase C18 Guard Column Kit, Upchurch Scientific®
Supplier: IDEX HEALTH & SCIENCE LLC
C-752
The guard column cartridges in this kit have bodies manufactured from PEEK* polymer and use biocompatible titanium frits on each end
The stainless steel high-pressure cartridge holder has PEEK inserts, preventing mobile phase contact with steel. This guard column will extend the life of columns and help ensure reproducible results.
ORDER
Expand All / Collapse All
Column Format I.D. x L Supplier No. VWR Catalog Number Unit Price Quantity
Analytical 4.3 x 10 mm C-752 23715-000 Each Retrieving
Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ACCESSORIES
Expand All / Collapse All
Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Guard Column Holder C-270 21511-032 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty