VWR® Critical Swab® Swab, Small Foam Head
Supplier: Avantor
VWR® Critical Swab®
TX818V
89022-994CS 335.28 USD
89022-994
VWR® Critical Swab® Swab, Small Foam Head
Swabs Swabs for Sampling
Swab features 100ppi open cell polyurethane foam with 100% virgin polypropylene handle.
  • Polyurethane foam head
  • Virgin polypropylene handle
  • No adhesive
Swab head is thermally bonded to the handle using no adhesive.