VWR® Critical Swab® Swab, Large Flexible Foam Head
TX814V
89022-988CS 349.81 USD
89022-988
VWR® Critical Swab® Swab, Large Flexible Foam Head
Swab features 100ppi closed cell polyurethane foam with a polypropylene/nylon handle.
  • Polyurethane foam head
  • Polypropylene/nylon handle
  • Augmented with adhesive
Swab head is thermally bonded to the handle and augmented with adhesive.