SP Wilmad-LabGlass Two-Piece Vacuum Traps with Straight Top Stem, SP Industries
Supplier: SP Industries
LG-11020-100 LG-11020-098 LG-11020-106 LG-11020-102
14203-556EA 277.68 USD
14203-556 14209-882 14209-794 14203-174
SP Wilmad-LabGlass Two-Piece Vacuum Traps with Straight Top Stem, SP Industries
Vacuum Traps
Made of borosilicate glass, these vacuum traps consist of two pieces with a straight top stem.
  • Body and stopper are connected by a full length Standard Taper joint
Both inlet and outlet stems are integral with the stopper.