Fiber-Lite® Illumination System 190-1, Dolan-Jenner
190-1
41446-076EA 996.45 USD
41446-076
Fiber-Lite® Illumination System 190-1, Dolan-Jenner
This system can be used independently for incident illumination from any angle or interface directly to stereo microscope ports for both incident and transmitted illumination.
Illuminator features a rugged convection-cooled enclosure to eliminate noise, vibration, and air particle generation. The three-position light intensity switch allows the selection of appropriate light levels and significantly increases lamp life at lower settings.
Ordering information: Supplied with one-year warranty on fiber optics. Complete with illuminator, single gooseneck light guide, focusing lens, and 30W halogen bulb. For additional accessories, see 41446-140 series.