Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Animal Sling Suits, Antron Engineering

Supplier: Antron Engineering

Sling suits are used with sling frames (see 50000-366 and -364 series) to support animals in an immobile position. They are made from durable, open mesh nylon.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Description Size Weight Range Supplier No. VWR Catalog Number Unit Price Quantity
Cat Sling Suit Medium 2.5–4.5 kg (51/2–10 lbs.) PRO-CATS2 50000-382 Each Retrieving Restricted
Cat Sling Suit Small up to 2.5 kg (51/2 lbs.) PRO-CATS1 50000-380 Each Retrieving Restricted
Cat Sling Suit Large 4.5 kg (10 lbs.) PRO-CATS3 50000-384 Each Retrieving Restricted
Dog Sling Suit X-Small 2.5–5 kg (51/2–11 lbs.) PRO-DS1 50000-388 Each Retrieving Restricted
Dog Sling Suit Large 13–18 kg (282/3–395/7 lbs.) PRO-DS4 50000-394 Each Retrieving Restricted
Dog Sling Suit Small 5–7 kg (11–152/5 lbs.) PRO-DS2 50000-390 Each Retrieving Restricted
Dog Sling Suit Medium 8–12 kg (173/5–261/2 lbs.) PRO-DS3 50000-392 Each Retrieving Restricted
Dog Sling Suit X-Large 18–25 kg (395/7–551/9 lbs.) PRO-DS5 50000-396 Each Retrieving Restricted
Rabbit Sling Suit X-Large 5.4 kg (12 lbs.) PRO-RAS5 50000-376 Each Retrieving Restricted
Rabbit Sling Suit Medium 3.6–4.5 kg (8–10 lbs.) PRO-RAS3 50000-372 Each Retrieving Restricted
Rabbit Sling Suit Small 2.7–3.6 kg (6–8 lbs.) PRO-RAS2 50000-370 Each Retrieving Restricted
Rabbit Sling Suit X-Small up to 2.7 kg (6 lbs.) PRO-RAS1 50000-368 Each Retrieving Restricted
Rabbit Sling Suit Large 4.5–5.4 kg (10–12 lbs.) PRO-RAS4 50000-374 Each Retrieving Restricted
Rodent Sling Suit Large 350–500 g (121/3–173/5 oz.) PRO-ROSS4 50000-360 Each Retrieving Restricted
Rodent Sling Suit Medium 250–350 g (84/5–121/3 oz.) PRO-ROSS3 50000-358 Each Retrieving Restricted
Rodent Sling Suit X-Small 100–150 g (31/2–52/7 oz.) PRO-ROSS1 50000-354 Each Retrieving Restricted
Rodent Sling Suit Small 150–250 g (52/7–84/5 oz.) PRO-ROSS2 50000-356 Each Retrieving Restricted
Rodent Sling Suit X-Large 500 g (173/5 oz.) PRO-ROSS5 50000-362 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty