Bunny Snuggles, Antron Engineering
Supplier: Antron Engineering
PRO-BS1 PRO-BS2 PRO-BS3
50000-234EA 276.56 USD
50000-234 50000-236 50000-238
Bunny Snuggles, Antron Engineering
Animal Supplies Animal Restraints
Bunny snuggles are ideal for short tests that require rabbit handling
Unlike other restraining methods, snuggles allow the handler to hold the subject while performing the procedure and do not immobilize the subject. This helps to reduce rabbit stress and discomfort, resulting in more efficient testing. The multiple flaps can be adjusted around the rabbit for effective yet comfortable restraint and vision restriction. Flaps may be opened or closed easily depending on the procedure needs. The bunny snuggle is made of nylon-coated canvas. Velcro® fasteners allow for easy adjustment between subjects.