Vacuum Trap, Twin Chamber, Ace Glass
Supplier: Ace Glass
8749-20
89063-892EA 624.22 USD
89063-892
Vacuum Trap, Twin Chamber, Ace Glass
Vacuum Traps
Twin-chamber vacuum trap provides double protection against undesired vapors in a vacuum system
Can be used in a 2L Dewar flask. Distance from outside of one body to outside of other is approximately 97mm. Inlet and outlet have serrated fittings with a 14–16mm O.D