LABOPORT® PowerDry® Self-Cleaning Vacuum Pumps, KNF Neuberger
Supplier: KNF
LABOPORT® PowerDry®
N860.3FT.40P N820.3FT.40P N842.3FT.40P N840.3FT.40P
46620-634EA 6493 USD
46620-634 46620-630 46620-636 46620-632
LABOPORT® PowerDry® Self-Cleaning Vacuum Pumps, KNF Neuberger
Pumps Vacuum Pumps
These pumps are ideal for use in rotary evaporation, distillation, vacuum oven drying, vacuum concentration, and gel drying
Pumps with a 20L/min. flow rate are ideal for gel drying. Pumps with a 34L/min. flow rate are ideal for vacuum concentration. The 60L/min. pumps are ideal for large scale vacuum concentration.
PowerDry® pumps instantly purge condensed vapors and restore end vacuum performance while running. An adjustable electronic timer automatically cycles every few minutes, removing condensed vapors from the pump without disturbing the vacuum process. Keeping the vacuum pump clean and dry leads to shorter processing times, lower maintenance costs, and less downtime.
The 400mL dry ice trap (26678-056) has a glass body and a Plexiglas® lid. It is recommended as a pre-trap to collect liquids and reduce processing time when used with pumps in rotary evaporation, distillation, gel drying, vacuum concentration, and vacuum ovens. For maximum efficiency, the dry ice trap should be placed before the pump.
Corrosion-resistant wetted parts made from fluoropolymer resin and Kalrez® resin make these pumps appropriate for use with even the most aggressive solvents.