Ubbelohde Viscometers, Calibrated, Technical Glass Products
Supplier: Q GLASS II LLC
UBB-2C UBB-1C UBB-1B UBB-0B UBB-4 UBB-5 UBB-3C UBB-3 UBB-2 UBB-4B UBB-2B UBB-1 UBB-3B UBB-0 UBB-0C UBB-4C
52945-884EA 335.7 USD InStock
52945-884 52945-829 52945-840 52945-809 52945-945 52945-964 52945-942 52945-920 52945-862 52945-947 52945-909 52945-807 52945-922 52945-805 52945-806 52945-949
Ubbelohde Viscometers, Calibrated, Technical Glass Products
Viscometers Capillary Viscometers
For measurement of kinematic viscosity of transparent liquids as described in ASTM Standard D445
Viscometer suspends through a 5.1cm (2") hole in the cover of a constant temperature bath by means of a round holder.
Standard viscometers in the petroleum industry. Ubbelohde viscometer constant does not vary with bath temperature, making it well suited for use at very high and low temperatures. These viscometers require a minimum sample of 11mL and liquid bath depth of 24.1cm (91/2"). Specifications conform to ASTM Standard D446. Calibrated by manufacturer and supplied with certificate.