Cross-Arm Viscometers, Calibrated, Technical Glass Products
Supplier: Technical Glass Products (TGP)
XA-4 XA-9 XA-3 XA-8 XA-10 XA-6 XA-2 XA-7 XA-5 XA-1
52946-264EA 474.14 USD
52946-264 52946-366 52946-242 52946-344 52946-388 52946-300 52946-220 52946-322 52946-286 52946-209
Cross-Arm Viscometers, Calibrated, Technical Glass Products
Viscometers Capillary Viscometers
Measure kinematic viscosity of transparent and opaque liquids by ASTM Standard D445
Also measure viscosity of cut-back asphalts and road oils at 60°C (140°F) and asphalt cements at 135°C (275°F), in the range of 30–6000 centistokes by ASTM D2170. Especially useful for limited sample sizes; require a minimum sample of 3mL. Can be filled and cleaned without removal from constant temperature bath. Liquid bath depth: 230mm. Specifications conform to ASTM Standard D446. Calibrated by manufacturer and supplied with certificate.
Accessories information: Rubber holder is drilled and split to accommodate the viscometer. Chemically-resistant plastic holder slips over viscometer and holds it firmly in place with a spring clip.
Ordering information: Holders are sold separately. A metal holder is also available; contact your VWR representative for more information.