VWR® Digitrate® Bottle-Top Burets
Supplier: VWR, part of Avantor
613-5287
75856-478EA 1532.46 USD
75856-478
VWR® Digitrate® Bottle-Top Burets
Digitrate bottle-top burets provide convenient bottle-top digital titration of any reagent.
  • Convenient bottle-top digital titration of any reagent
  • Constructed of borosilicate glass
  • Fluoropolymer resin components
  • 0.2% accuracy
  • 0.1% reproducibility
Burets feature large LCD display, safety dispense nozzle protector, and “zero” reset button for multiple titrations
Amber, low-profile bottle (sold separately) has a side opening which eliminates the need to remove the Digitrate from the bottle during filling. A safety funnel (also sold separately) has an off-set stem for safer filling of the side neck on the bottle.
Simple user calibration procedure. Precision valve mechanism ensures easy priming with no leakage back into the reservoir. Chemically-resistant liquid pathway. Visual indication of last calibration. Head rotates 360° for use in any position. Burets may be easily dismantled for cleaning without the use of tools. Simple removal of electronics for autoclaving. Digitrate fits directly onto 30 mm neck reservoirs. Each instrument is individually calibrated against National Institute of Standards and Technology Traceable equipment.
Ordering information: Supplied with 33, 38, and 45 mm bottle neck adapters and a calibration certificate. Also supplied with manufacturer’s three-year warranty. A bottle is not supplied; amber squat bottles with safety side filler necks are sold separately.