VWR® Traceable® Sentry Minimum/Maximum Memory Thermometers
Supplier: VWR, part of Avantor
VC1057 4121 VC4322 VC2057 9657
61161-289EA 30.36 USD
61161-289 10171-316 10171-596 15551-284 10171-480
VWR® Traceable® Sentry Minimum/Maximum Memory Thermometers
Monitor temperature of any experiment even when not there.
  • Traceable
  • Triple display design
  • Epoxy tip sensor
  • Simple one-button operation
Triple display simultaneously shows current, high, and low temperatures
Probe with epoxy tip sensor and 3m (10') cable may be placed underwater, or in refrigerators and freezers. Thermometer is available with bottle probe filled with nontoxic glycol to simulate liquid temperatures in freezers and refrigerators. Cable diameter of 1.6mm (1/16") allows refrigerator door to close on it with no effect. Low battery is signaled on the display. Simple one-button operation resets the minimum/maximum memories. High-impact ABS plastic case has a flip-open stand, plastic hanging hook, magnet, and Velcro® mounting tape.
Vaccine thermometers 89094-788 and 89094-790 have a sensor probe that is sealed in a 5 mL bottle filled with GRAS nontoxic glycol. The bottle mimics vaccine temperature and fits in a vaccine tray.
Instantly shows if water bath, freezer, or incubator is within temperature specifications.
Certifications: Thermometers fulfill all CDC thermometer and vaccine thermometer requirements. A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with probe and cable, wire mounting bracket for easy sensor placement, and a 1.5V silver-oxide battery for continuous one-year use.