VWR® Screw-Thread Vials 10-425
Supplier: VWR
VWR®
VWC4010-1W VWC4010-1 VWC4010-2W
82028-428CS 466.63 USD
82028-428 82028-432 82028-426
VWR® Screw-Thread Vials 10-425
Vials
Wide opening screw-thread vials are manufactured from borosilicate glass and have a maximum fill volume of 1.8 ml.
  • Borosilicate glass
  • Uniformly flat bottoms
  • Wide neck is easy to fill
Clear vials are made from 33 expansion borosilicate glass. Amber vials are made from 51A amber borosilicate glass.
Ordering information: Screw caps with preattached septa are available separately.