Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Kahnetics® Stainless Steel Needles, Cooper Tools

Supplier: APEX TOOL GROUP
Kahnetics®
KDS1712P KDS1912P KDS181P KDS2512P KDS2312P KDS2212P KDS201P KDS16112P KDS141P KDS161P KDS1512P KDS1412P KDS221P
Blunt needles have plastic, color-coded hubs

All needles lock onto Luer-lock style syringes. Available in lengths from 12.7 to 38.1mm (1/2 to 11/2"). Disposable. Sold in packs of 50.



 

ORDER

Expand All / Collapse All

12.7 mm (1/2") Length
Gauge Inner Diameter O.D. Color Supplier No. VWR Catalog Number Unit Price Quantity
14 1.70 mm (0.067") 2.11 mm (0.083") Dark Green KDS1412P 37696-944 Each Retrieving
15 1.52 mm (0.060") 1.83 mm (0.072") Orange KDS1512P 37696-974 Each Retrieving
17 1.14 mm (0.045") 1.52 mm (0.060") White KDS1712P 37695-010 Each Retrieving
19 0.81 mm (0.032") 1.07 mm (0.042") Brown KDS1912P 37695-054 Each Retrieving
22 0.48 mm (0.019") 0.71 mm (0.028") Black KDS2212P 37695-140 Each Retrieving
23 0.43 mm (0.017") 0.64 mm (0.025") Light Blue KDS2312P 37695-168 Each Retrieving
25 0.30 mm (0.012") 0.51 mm (0.020") Blue KDS2512P 37695-196 Each Retrieving

25.4 mm (1") Length
Gauge Inner Diameter O.D. Color Supplier No. VWR Catalog Number Unit Price Quantity
14 1.70 mm (0.067") 2.11 mm (0.083") Dark Green KDS141P 37696-942 Each Retrieving
16 1.35 mm (0.053") 1.63 mm (0.064") Purple KDS161P 37696-990 Each Retrieving
18 0.97 mm (0.038") 1.27 mm (0.050") Pink KDS181P 37695-022 Each Retrieving
20 0.66 mm (0.026") 0.89 mm (0.035") Yellow KDS201P 37695-084 Each Retrieving
22 0.48 mm (0.019") 0.71 mm (0.028") Black KDS221P 37695-138 Each Retrieving

38.1 mm (11/2") Length
Gauge Inner Diameter O.D. Color Supplier No. VWR Catalog Number Unit Price Quantity
16 1.35 mm (0.053") 1.63 mm (0.064") Purple KDS16112P 37696-986 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty