Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
You Searched for: "Picric acid saturated solution in water"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  Ricca Chemical
Description:   2,4,6-Trinitrophenol. CAS RN 88-89-1. Formula Weight: 229.10. Saturated aqueous solution, approximately 1.2% (w/v). Packaged in a plastic container.Subject to postal/shipping restrictions. 1L.
SDS
Small Business Enterprise