DuPont™ ProClean® Sleeves
PC500SWH000B PC500SWH000B
13452-572CS 226.54 USD
CA13452-572 13452-572
DuPont™ ProClean® Sleeves
ProClean® offers improved non-hazardous light liquid splash protection.
  • Serged seams
  • Covered elastic opening at both ends
  • One size fits most
ProClean® is made from a microporous composite fabric. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Elastic openings at bicep and wrist ensure secure fit and protection.
These sleeves feature elastic at both biceps, measuring 73/4", and wrist, measuring 51/4".
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.