Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SMS Coveralls, International ENVIROGUARD™

Supplier: INTERNATIONAL ENVIROGUARD
2229XXXXL 2229BXXXL 2229XL 2229BXXXXL 2229BXXL 2229BM 2229BXL 2229XXXL 2229BL 2229L 2229M
Coveralls are constructed of three-layer, spunbonded-meltblown-spunbonded material, which offers splash resistance yet is cool and comfortable to wear

Garments may be used for pharmaceutical and general industrial applications, as well as manufacturing, asbestos removal, and assembly operations. Coveralls feature a zipper front, hood, boots, and elastic wrists, ankles, and back. Fabric weight: 60g.




 

ORDER

Expand All / Collapse All
Size Color Supplier No. VWR Catalog Number Unit Price Quantity
3X-Large White 2229XXXL 83007-092 Case of 25 Retrieving
3X-Large Denim Blue 2229BXXXL 83007-186 Case of 25 Retrieving
4X-Large White 2229XXXXL 83007-094 Case of 25 Retrieving
4X-Large Denim Blue 2229BXXXXL 83007-188 Case of 25 Retrieving
Large Denim Blue 2229BL 83007-180 Case of 25 Retrieving
Large White 2229L 83007-086 Case of 25 Retrieving
Medium White 2229M 83007-084 Case of 25 Retrieving
Medium Denim Blue 2229BM 83007-178 Case of 25 Retrieving
X-Large White 2229XL 83007-088 Case of 25 Retrieving
X-Large Denim Blue 2229BXL 83007-182 Case of 25 Retrieving
XX-Large Denim Blue 2229BXXL 83007-184 Case of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty