KIMAX® Medium Length West Condenser, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
18185-1420
89001-660CS 327.32 USD
89001-660
KIMAX® Medium Length West Condenser, Kimble Chase
Condensers
KIMAX® 200mm (77/8") one-piece sealed unit with a [ST] 14/20 outer joint at the inlet and a [ST] 14/20 inner joint at the outlet
Inlet end is sloped to avoid trapping condensate. Inner tube is light-walled with a small diameter jacket to keep minimal space between them. Hose connections accept 7.9mm (5/16") I.D. flexible tubing. (Kimble Chase 18185 1420)