Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DuPont™ Tychem® 9000 Coveralls with Respirator Hood

Supplier: DuPont
Tychem®
BR127TYLMD00 BR127TYLSM00 BR127TYLXL0002BN BR127TYLLG00 BR127TYL5X000200 BR127TYLMD0002BN BR127TYL4X000200 BR127TYL6X000200 BR127TYL4X00 BR127TYLXL00 BR127TYL3X000200 BR127TYLSM0002BN BR127TYL3X00 BR127TYL2X00 BR127TYLLG000200 BR127TYL5X0002BN BR127TYL4X0002BN BR127TYL3X0002BN BR127TYL2X000200 BR127TYLXL000200 BR127TYLMD000200 BR127TYLLG0002BN BR127TYLSM000200
DuPont™ Tychem® 9000 offers wide-ranging protection backed by extensive fabric permeation barrier testing.

  • Respirator fit hood
  • Elastic wrists and ankles
  • Storm flap with adhesive closure
  • Taped seams

Tychem® 9000 fabric provides at least 30 minutes of protection against more than 280 chemical challenges, including chemical warfare agents. Tychem® 9000 offers tear, puncture and abrasion resistant fabrics for lasting protection. Tychem® 9000 garments are designed for enhanced comfort and ease of movement. These coveralls are high-visibility yellow.

Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Three-piece hood design with longer zipper that extends to chin for complete coverage of neck area and good fit around a respirator. Coveralls feature a strong zipper for tough, flexible closure. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Elastic openings at wrist and ankle ensure a tighter fit.

Certifications: Thickness meets ASTM D1777.




 

ORDER

Expand All / Collapse All

Tychem® 9000 Coveralls
Size Chest Size Color Inseam Sleeve Supplier No. VWR Catalog Number Unit Price Quantity
S 233/8" Yellow 28" 323/4" BR127TYLSM000200 10010-280 Case of 2 Retrieving
M 247/8" Yellow 281/2" 34" BR127TYLMD000200 10010-282 Case of 2 Retrieving
L 263/8" Yellow 291/2" 351/4" BR127TYLLG000200 10010-284 Case of 2 Retrieving
XL 277/8" Yellow 297/8" 363/4" BR127TYLXL000200 10010-286 Case of 2 Retrieving
2X 29" Yellow 307/8" 377/8" BR127TYL2X000200 10010-288 Case of 2 Retrieving
3X 31" Yellow 317/8" 39" BR127TYL3X000200 10010-290 Case of 2 Retrieving
4X 325/8" Yellow 325/8" 40" BR127TYL4X000200 10010-292 Case of 2 Retrieving
5X 34" Yellow 337/8" 407/8" BR127TYL5X000200 76187-184 Case of 2 Retrieving
6X 353/4" Yellow 35" 413/4" BR127TYL6X000200 76187-188 Case of 2 Retrieving

Tychem® 9000 Coveralls, Berry Amendment Compliant
Size Chest Size Color Inseam Sleeve Supplier No. VWR Catalog Number Unit Price Quantity
S 233/8" Yellow 28" 323/4" BR127TYLSM0002BN 76187-194 Case of 2 Retrieving
M 247/8" Yellow 281/2" 34" BR127TYLMD0002BN 76187-192 Case of 2 Retrieving
L 263/8" Yellow 291/2" 351/4" BR127TYLLG0002BN 76187-190 Case of 2 Retrieving
XL 277/8" Yellow 297/8" 363/4" BR127TYLXL0002BN 76187-196 Case of 2 Retrieving
3X 31" Yellow 317/8" 39" BR127TYL3X0002BN 76187-180 Case of 2 Retrieving
4X 325/8" Yellow 325/8" 40" BR127TYL4X0002BN 76187-182 Case of 2 Retrieving
5X 34" Yellow 337/8" 407/8" BR127TYL5X0002BN 76187-186 Case of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

COMPLEMENTARY PRODUCTS

Expand All / Collapse All

Complementary Tychem® 9000 Hood
Length Width Supplier No. VWR Catalog Number Unit Price Quantity
213/4" 293/4" BR651TYL00000200 76187-162 Case of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping