VWR® Critical Cover® AlphaGuard® Coveralls
Supplier: VWR, part of Avantor
CV-74032-4
14001-582CS 419.31 USD
14001-582
VWR® Critical Cover® AlphaGuard® Coveralls
The unique multilayered construction of these coveralls acts as a fluid and particulate barrier that remains soft, comfortable, and breathable.
  • Soft, breathable, comfortable material
  • No pocket
  • Serged seams
AlphaGuard® material features an antistatic treatment to reduce static charges. Coveralls feature inset sleeves, a front zipper, and elastic cuffs, ankles, and back.