Would you like to log in again?
Portable Laboratory Micro-Burner, Global Market Connections
Supplier: GLOBAL MARKET CONNECTIONS, LTD
MF-2001
Compact and durable, this micro-burner features push-button ignition with adjustable flame control, as well as a tip-resistant design with an easy-grip handle
Operation time exceeds one hour. Refills quickly and easily with standard butane fuel. This model requires butane fuel cylinder. No wicks to replace or gas lines to install. An optional beaker stand, constructed of high-quality gray powder-coating on cast iron components with three chrome-plated legs, supports beaker sizes of up to 1000 mL while resisting corrosion and discoloration. The optional crucible cradle allows safe heating, cooling, and weighing of the crucible and its contents.
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Portable Laboratory Micro-Burner MF-2001 89025-954 Each Retrieving
Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ACCESSORIES
Expand All / Collapse All
Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Beaker Stand BS-2001 89025-948 Each Retrieving
Pack of 12 Retrieving
Crucible Cradle CC-2006 89025-952 Each Retrieving
Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty