Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Accessories

Supplier: Expedeon
VGS2020R-177 VGE1020-177 VGS2020RC177 VGS1020R-177 VGE2020-177 GPC-0002-177 GPC-0001-177 VGS1520R-177 VGE0520-177 VGS7520R-177 VGE1520-177 VGE7520-177 NGP-17720-NR NGP-17720-N VGS1520RC177 VGS7520RC177 VGS1020RC177
Accessories for DGGE systems (CBDGGE-1001 series).




 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Clamp, white, jaws closed in resting position GPC-0002-177 CBGPC-0002-177 Each Retrieving Restricted
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Clamp, white, jaws slightly open in resting position GPC-0001-177 CBGPC-0001-177 Each Retrieving Restricted
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Gel Wrap Gasket, 0.75 mm thick x 22 cm length VGE7520-177 CBVGE7520-177 Each Retrieving Restricted
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Gel Wrap Gasket, 1.0 mm thick x 22 cm length VGE1020-177 CBVGE1020-177 Each Retrieving Restricted
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Gel Wrap Gasket, 1.5 mm thick x 22 cm length VGE1520-177 CBVGE1520-177 Each Retrieving Restricted
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Glass plate set, 7.7 cm width x 22 cm length NGP-17720-N 71003-642 Each Retrieving
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Spacer Sets for perpendicular DGGE, 0.75 mm thick x 22 cm length VGS7520RC177 CBVGS7520RC177 Each Retrieving Restricted
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Spacer Sets for perpendicular DGGE, 1.0 mm thick x 22 cm length VGS1020RC177 CBVGS1020RC177 Each Retrieving Restricted
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Spacer Sets for perpendicular DGGE, 1.5 mm thick x 22 cm length VGS1520RC177 CBVGS1520RC177 Each Retrieving Restricted
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Spacer Sets for vertical DGGE, 0.75 mm thick x 22 cm length VGS7520R-177 CBVGS7520R-177 Each Retrieving Restricted
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Spacer Sets for vertical DGGE, 1.0 mm thick x 22 cm length VGS1020R-177 CBVGS1020R-177 Each Retrieving Restricted
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Spacer Sets for vertical DGGE, 1.5 mm thick x 22 cm length VGS1520R-177 CBVGS1520R-177 Each Retrieving Restricted
Minority or Woman-Owned Business Enterprise Small Business Enterprise
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty