Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Breathe Easy™ Powered Air Purifying Respirator (PAPR) System, 3M™

Supplier: 3M OCCUPATIONAL HEALTH
Breath Easy™
520-15-00 520-17-00
PAPR system provides comfortable and efficient powered air protection

Assemblies include motor/blower, battery pack, belt, and flowmeter. Assembly 56221-894 is “intrinsically safe” (i.e., incapable of creating a spark or explosion). Assemblies use interchangeable cartridges/filters that protect against gases, vapors, or particulates.

Headpieces are constructed of Tychem® QC for protection from chemicals; they are available in hood, sealed-seam hood, or head cover styles. The loose-fitting hood and sealed-seem hood accommodate facial hair and glasses. The lightweight head cover is available in white or blue. Sealed-seam hood assembly 67101-162 features a built-in collar and cap suspension. Sealed-seam hood assembly 67101-164 features a built-in collar and hardhat.

Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.

Ambient air is drawn through the filter cartridge, supplying filtered air through the breathing tube to the headpiece.

Certifications: NIOSH approved.

Ordering information: PAPR assembly, headpiece, smart charger, filters, and breathing tube are ordered separately.



 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Breathe Easy™ “Intrinsically Safe” Turbo PAPR Assembly 520-17-00 56221-894 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
84L cm (33") Breathing Tube for Head Covers 520-03-32R01 67100-276 Each Retrieving
91L cm (36") Breathing Tube for Hoods 520-01-00R01 67100-204 Each Retrieving
Acid Gas/HEPA Filter for Breathe Easy™ Turbo PAPR Assembly 453-07-01R06 67100-144 Pack of 6 Retrieving
Ammonia/HEPA Filter for Breathe Easy™ Turbo PAPR Assembly 453-02-01R06 67100-140 Pack of 6 Retrieving
Formaldehyde/HEPA Filter for Breathe Easy™ Turbo PAPR Assembly 453-01-01R06 67100-138 Pack of 6 Retrieving
HEPA Filter for Breathe Easy™ “Intrinsically Safe” Turbo PAPR Assembly 450-00-01R12 67100-126 Case of 12 Retrieving
Pack of 3 Retrieving
Large Size Head Cover, White BE-12L-3 67100-336 Pack of 3 Retrieving
Large Size Hood, White BE-10L-3 67100-350 Pack of 3 Retrieving
Organic Vapor/Chlorine/Hydrogen Chloride/Sulfur Dioxide/High Efficiency Cartridge Breathe Easy™ Turbo PAPR Assembly 453-03-01R06 67100-141 Case of 6 Retrieving
Pack of 3 Retrieving
Organic Vapor/HEPA Filter for Breathe Easy™ Turbo PAPR Assembly 453-00-01R06 67100-135 Case of 6 Retrieving
Each Retrieving
Regular Size Head Cover, White BE-12-3 67100-334 Pack of 3 Retrieving
Regular Size Hood, White BE-10-3 67100-302 Pack of 3 Retrieving
Regular Size Sealed-Seam Hood Assembly with Collar and Cap Suspension H-611 67101-162 Case of 1 Retrieving
Regular Size Sealed-Seam Hood with Collar H-610-5 67100-856 Case of 5 Retrieving
Pack of 1 Retrieving
Smart Battery Charger with LED Status Indicator, 5-Unit 520-03-72 67100-286 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty