ValueCare® Half-Back Isolation Gowns, TECHstyles®
4540-150 4540-145
10828-756CS 156.15 USD
10828-756 10828-758
ValueCare® Half-Back Isolation Gowns, TECHstyles®
These disposable, latex-free protective gowns are functional and economical
  • Sewn gown
  • Tie neck, waist tie
  • Half-back
  • Available with either elastic or knit cuffs
Constructed from polyethylene-coated spunbonded polypropylene, the gowns offer a high level of fluid resistance. These gowns are designed for high-risk exposure in settings where there is probable occurrence of spraying or splashing of fluids.
Packaging: Packaged 10 per bag, 5 bags per case.