KLEENGUARD® A60 Bloodborne Pathogen and Chemical Splash Protection Hood, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
45343-10 45343
82004-030CS 256.31 USD
82004-030 CA82004-030
KLEENGUARD® A60 Bloodborne Pathogen and Chemical Splash Protection Hood, KIMBERLY-CLARK PROFESSIONAL®
Ideal hood for wet jobs
Three-layer fabric construction features a middle layer of microporous film that allows heat and sweat vapor to escape, yet protects against many dry particulates and liquids — even under pressure. Thicker, more uniform outer layers provide protection against tearing and abrasion. Fabric passes ASTM F1670/1671 testing for penetration of blood, body fluids, and bloodborne pathogens. Elastic face opening