KIMBLE® KONTES® Microscale Vacuum Adapter, GPI to [ST], DWK Life Sciences
Supplier: DWK Life Sciences
KONTES®
746040-0000
KT746040-0000CS 129.58 USD
KT746040-0000
KIMBLE® KONTES® Microscale Vacuum Adapter, GPI to [ST], DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
These microscale vacuum GPI to standard taper adapters are ideal for closed system applications.
  • Top GPI thread
  • Bottom [ST] joint
  • 105° angle bend
  • With hose connector and drip tip
Adapter features a 20-400 GPI thread on the top and a [ST] 14/10 joint on the bottom. Bent at 105° with a #1 hose connector and a drip tip.