HandleLock® B-Jacket Bottle Jacket, Honeywell Burdick & Jackson®
Supplier: Honeywell Research Chemicals
HandleLock®
S1337-20 S1337-20
BJ-S1337-21CS 413.6 USD
BJ-S1337-21 CABJ-S1337-20
HandleLock® B-Jacket Bottle Jacket, Honeywell Burdick & Jackson®
Carriers Bottle Carriers/Containers
Resusable bottle jacket provides virtually unbreakable, secondary containment for storage and handling of 4L solvent bottles as recommended by OSHA Standard 29 CFR 1910.1450 Appendix A
These bottle jacket are made of polyethylene.
Contains spills and broken glass in the event a bottle is dropped or broken. Features rugged polymer construction with convenient HandleLock® mechanism. Solvent labels are visible through the bottle jacket.