Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dog Jackets, Antron Engineering

Supplier: Antron Engineering

Full-length dog jackets are suitable for a variety of applications including intravenous infusion, physiological investigations, and protecting surgical sites. Jackets are made of durable, open mesh nylon for a comfortable fit. They feature two zipper closures and one spandex expansion panel. Pockets, also made from open mesh nylon, are available to accommodate portable equipment.

Ordering Information: Contact your VWR sales representative for information on additional spandex expansion panels or jackets in a 3/4-length style. Pockets are custom sewn on jackets; specify choice of pocket and desired placement when ordering.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Size Weight Range Supplier No. VWR Catalog Number Unit Price Quantity
Large 13–18 kg (282/3–395/7 lbs.) PRO-DOJ4 50000-246 Each Retrieving Restricted
Medium 8–12 kg (173/5–261/2 lbs.) PRO-DOJ3 50000-244 Each Retrieving Restricted
Small 5–7 kg (11–152/5 lbs.) PRO-DOJ2 50000-242 Each Retrieving Restricted
X-Large 18–25 kg (395/7–551/9 lbs.) PRO-DOJ5 50000-248 Each Retrieving Restricted
X-Small 2.5–5 kg (51/2–11 lbs.) PRO-DOJ1 50000-240 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories for Dog Jackets, Antron Engineering
Description Dimensions Supplier No. VWR Catalog Number Unit Price Quantity
Jacket Pocket with 17.8 cm (7") Top Zipper 7.6W x 17.8L x 5.1D cm (3 x 7 x 2") PRO-JP1 50000-290 Each Retrieving Restricted
Jacket Pocket with 17.8 cm (7") Top Zipper 15.2W x 10.2L x 5.1D cm (6 x 4 x 2") PRO-JP4 50000-294 Each Retrieving Restricted
Jacket Pocket with 22.9 cm (9") Side Zipper 13.3W x 8.9L x 3.8D cm (51/4 x 31/2 x 11/2") PRO-JP5 50000-296 Each Retrieving Restricted
Jacket Pocket with 27.9 cm (11") Side Zipper 17.8W x 14L x 3.8D cm (7 x 51/2 x 11/2") PRO-JP3 50000-292 Each Retrieving Restricted
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty