SP Bel-Art Economy Nonsterile Sample Spoons, Bel-Art Products, a part of SP
Supplier: Bel-Art
Sterileware®
F36740-0024 F36740-0025 F36740-0023 F36740-0022 F36740-0021
74950-198EA 19.84 USD
74950-198 74950-196 74950-194 74950-192 74950-190
SP Bel-Art Economy Nonsterile Sample Spoons, Bel-Art Products, a part of SP
Spoons
These economical measuring devices are ideal for retrieving small, specific volume samples of powders or liquids.
  • Economical
  • Unique Flat-bottom shape
  • Nonsterile
  • White FDA-grade polystyrene construction
Their unique flat-bottom shape allows them to be used as weighing boats for additional sampling accuracy.
Use a separate spoon for each material sampled to avoid cross contamination. All sizes are made from white, FDA-grade high-impact polystyrene plastic for durability and stiffness when sampling packed powders.
Ordering information: Supplied 12 per bag.