CFM Anemometer Data Logger, Sper Scientific
840001
14900-504EA 572.34 USD
14900-504
CFM Anemometer Data Logger, Sper Scientific
Automatically records up to 16,000 data points in the field along with time and date
Data points can later be downloaded to a computer via the RS-232 port and the supplied software. The software also enables real-time data logging directly from the meter to the computer. The microprocessor calculates air volume (CFM) as well as air velocity, and simultaneously displays air temperature in Celsius or Fahrenheit. Unit also reads Type-K or Type-J thermocouple probes and features a detachable probe, fold-out tripod, large easy-to-read LCD, and calendar, clock, auto power off, minimum/maximum display, and hold functions.
Ordering information: Supplied with protective foam-lined carrying case, software, RS-232 computer connection cable, four AA batteries, and a 3V button cell battery. Unit 14900-504 includes a metal vane anemometer probe. Unit 14900-506 includes telescopic hotwire anemometer probe.