Super Erecta® Reel Shelving, Super Erecta Brite® Finish, Metro™
Supplier: Metro International
Super Erecta™ Super Erecta®
R1836BR-13 R1836BR-13 R1836BR-7 R1836BR-7
22221-422EA 232.08 USD
CA22221-422 CA22221-420 22221-422 22221-420
Super Erecta® Reel Shelving, Super Erecta Brite® Finish, Metro™
Furniture Shelves
Shelves provide efficient storage and transport of standard component reels.
  • Super Erecta Brite® shelves are made of zinc-plated steel
  • Shelves adjust easily without the use of tools
SiteSelect® chrome-plated stationary and mobile posts allow shelves to be readjusted and snapped into place at precise 2.5 cm (1") increments along the entire height of the post. An easily identifiable double groove is positioned every 20.3 cm (8") to further aid shelf alignment. Stationary posts are fitted with adjustable leveling bolts for use on uneven surfaces. Mobile posts require the purchase of stem casters. Stem casters (see 19766-236 series) and shelf dividers and ledges (see 22233-212 series) are available separately.
Shelves can be mixed and matched with standard Super Erecta® shelves, stem casters, and split sleeves (see 19766-814 series) to create a system meeting the exacting needs of PCB manufacturers. Shelving can be made conductive for ESD safety when used with conductive split sleeves and conductive casters.
Note: The actual length of the shelves is 3.2 mm (¹/₈") shorter than the nominal dimension shown. Stationary posts over 189.6 cm high and mobile posts over 138.6 cm high are not for use with 35.6 cm wide shelves. The height of the chrome-plated and stainless steel stationary posts includes a leveling bolt and cap.
Certifications: Shelves meet MIL-S-40144E specifications.