Piperazine, anhydrous, OR, Macron Fine Chemicals™

Danger


Management of Change (MOC) category = N

MACR2675-59
MK267559EA 633.42 USD
MK267559
Piperazine, anhydrous, OR, Macron Fine Chemicals™
Piperazine

Macron Fine Chemicals™ OR: organic reagents that are suitable for research applications

Formula: C₄H₁₀N₂
MW: 86.14 g/mol
Boiling Pt: 146 °C (1013 hPa)
Melting Pt: 44…46 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00005953
CAS Number: 110-85-0
EINECS: 203-808-3
UN: 2579
ADR: 8,III
Merck Index: 12,07617

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Infrared Spectrum Passes Test
Melting Point 109 - 113 °C
Appearance (white flakes)
CAUTION! Material absorbs carbon dioxide from the air. Keep tightly closed.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR