Potassium hexacyanoferrate(II) trihydrate 98.5-102.0%, crystals, AR® ACS, Macron Fine Chemicals™

Synonyms: Tetrapotassium hexacyanoferrate trihydratePotassiumhexacyanoferrate(II) trihydratePotassium ferrocyanide trihydrate

6932-04 6932-05 MACR6932-04
MK6932-04CS 750.27 USD
CAMK693204 MK6932-04 MK693205
Potassium hexacyanoferrate(II) trihydrate 98.5-102.0%, crystals, AR® ACS, Macron Fine Chemicals™
Potassium hexacyanoferrate (II) trihydrate
Formula: K₄Fe(CN)₆·3H₂O
MW: 422.39 g/mol
Melting Pt: 70 °C
Density: 1.889 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167023
CAS Number: 14459-95-1
EINECS: 237-722-2
Merck Index: 12,07793

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (K₄Fe(CN)₆·3H₂O) 98.5 - 102.0 %
ACS - Appearance (pale yellow crystals) Passes Test
ACS - Chloride (Cl) ≤0.01 %
ACS - Insoluble Matter ≤0.005 %
ACS - Sulfate (SO₄) Passes Test
Solubility Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR