Petroleum spirit, 35…60 °C, UltimAR® for HPLC, for gas chromatography, for spectrophotometry, Macron Fine Chemicals™

Danger

Synonyms: Petroleum etherGasolinePetrol

Management of Change (MOC) category = NI

MACRH489-10 H489-06 MACRH489-06 H489-10
MKH48906CS 715.62 USD
MKH48906 CAMKH48910 CAMKH48906 MKH48910
Petroleum spirit, 35…60 °C, UltimAR® for HPLC, for gas chromatography, for spectrophotometry, Macron Fine Chemicals™
Petroleum spirit

UltimAR™ solvents let you consolidate the number of solvent grades used in a diverse variety of applications, saving money and storage space without compromising quality and reliability of test results. They’re tested for key analytical parameters in HPLC, GC and spectrophotometry.

Boiling Pt: 35…60 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Acidity Passes Test
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.0001 %
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.02 %
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
ACS - GC-FID Sensitive Impurities (as 2-Octanol) Single Peak (µg/L) ≤5
ACS - Sum of the Peaks (µg/L) ≤10
ACS - Neat Solvent Front Characterization Passes Test
ACS - Boiling Range(Lower) 35 - 60 °C
ACS - Boiling Range(Upper) 35 - 60 °C
Appearance (clear liquid free from suspended foreign matter or sediment)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR