You Searched For: 2-Chloro-5-(trifluoromethyl)pyridine
5  results were found
SearchResultCount:"5"
Sort Results
List View Easy View
Rate These Search Results
Supplier: TCI America
Description: CAS Number: 52334-81-3 MDL Number: MFCD00042225 Molecular Formula: C6H3ClF3N Molecular Weight: 181.54 Purity/Analysis Method: 97.0% (GC) Form...
Supplier: Thermo Scientific Chemicals
Description: . Grade:97, Melting Point C30-32*. Boiling Point C:152-153*. C6H3ClF3N. 52334-81-3. HARMFUL IRRITANT
Supplier: Thermo Scientific Chemicals
Description: 2-CHLORO5(TRIFLUOROMETHYL)PYRIDN98+% 25G
Supplier: Ambeed
Description: 2-Chloro-5-trifluoromethylpyridine, Purity: 98%, CAS Number: 52334-81-3, Appearance: Solid or semi-solid or liquid or lump, Storage: Keep in dark place, Sealed in dry, Room Temperature, Size: 500g
New Product
Supplier: Matrix Scientific
Description: MF=C6H3Clf3N MW=181.55 Cas=52334-81-3 MDL=MFCD00042225 100G
SDS
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 5 of 5
no targeter for Bottom