    Sort by:
  • Search Within Results
4-chloro-2-methylphenyl-isocyanate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   . Grade:98, Melting Point C42-43*. Boiling Point C:103*/13mm. C8H6ClNO. 37408-18-7. HIGHLY TOXIC IRRITANT SENSITISING KEEP COLD MOISTURE SENSITIVE