    Sort by:
  • Search Within Results
1-(3-Chloropropoxy)-4-fluorobenzene
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  TCI America
Description:   CAS Number: 1716-42-3 Molecular Formula: C9H10ClFO Molecular Weight: 188.63 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling poin...